missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12698-1-AP-150UL
This item is not returnable.
View return policy
Description
Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of KLK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.Specifications
| Kallikrein 11 | |
| Polyclonal | |
| Unconjugated | |
| KLK11 | |
| Hippostasin, hK11, Kallikrein 11, KLK11, PRSS20, Serine protease 20, TLSP, Trypsin like protease | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11012, 292849, 56538 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9QYN3, Q9UBX7 | |
| Klk11 | |
| Kallikrein 11 Fusion Protein Ag3427 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction