missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99572-50ul
This item is not returnable.
View return policy
Description
Kallikrein 11 Polyclonal antibody specifically detects Kallikrein 11 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Kallikrein 11 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:5000-1:10000 | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease | |
| Produced in rabbits immunized with purified, recombinant Human Kallikrein 11 (Accession#: NP_006844.1; Met 1-Asn 250) | |
| 50 μg | |
| Cancer | |
| 11012 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction