missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 11 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-429R
This item is not returnable.
View return policy
Description
Kallikrein 11 Polyclonal antibody specifically detects Kallikrein 11 in Human samples. It is validated for Western Blot
Specifications
| Kallikrein 11 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease | |
| Residues 68-82 [YIVHLGQHNLQKEEG] of the 35 kDa human hippostasin/KLK11protein. Sequence identity to mouse and rat: 86%. | |
| 0.1 mL | |
| Cancer | |
| 11012 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction