missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 11 Antibody (121), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18360380 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360380 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360380 Supplier Novus Biologicals Supplier No. NBP289630AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Kallikrein 11 Monoclonal specifically detects Kallikrein 11 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 11
Applications Western Blot, ELISA
Classification Monoclonal
Clone 121
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 11 (Met 1-Asn 250; NP_006844.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11012
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.