missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 11 Antibody (121), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89630AF594
This item is not returnable.
View return policy
Description
Kallikrein 11 Monoclonal specifically detects Kallikrein 11 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Kallikrein 11 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 11 (Met 1-Asn 250; NP_006844.1). | |
| 0.1 mL | |
| Cancer | |
| 11012 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 121 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction