missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NB100-57855

Product Code. 18007483

  • £397.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Kallikrein 1 Polyclonal specifically detects Kallikrein 1 in Human samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kallikrein 1
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, Klk6, KLKR, Tissue kallikrein
Goat
Affinity Purified
RUO
Primary
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Pig and Cow.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000
NP_002248.1
KLK1
Peptide with sequence PNDECKKAHVQK corresponding to internal region according to NP_002248.1.
0.1 mg
Cancer
3816
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only