missing translation for 'onlineSavingsMsg'
Learn More

Kalecide, TRC

Product Code. 30392863
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392863 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392863 Supplier Toronto Research Chemicals Supplier No. TRCK4450151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24738-51-0
Molecular Formula C16 H29 N O
Quantity 1 mg
Synonym (2E,4E)-N-(2-methylpropyl)-2,4-dodecadienamide, Kalecide
InChI Formula InChI=1S/C16H29NO/c1-4-5-6-7-8-9-10-11-12-13-16(18)17-14-15(2)3/h10-13,15H,4-9,14H2,1-3H3,(H,17,18)/b11-10+,13-12+
SMILES CCCCCCC\C=C\C=C\C(=O)NCC(C)C
IUPAC Name (2E,4E)-N-isobutyldodeca-2,4-dienamide
Molecular Weight (g/mol) 251.41
Formula Weight 251.2249
Chemical Name or Material Dodeca-2(E),4(E)-Dienoic Acid Isobutylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.