missing translation for 'onlineSavingsMsg'
Learn More

KAL1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18412358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412358 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412358 Supplier Novus Biologicals Supplier No. NBP176580JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KAL1 Polyclonal antibody specifically detects KAL1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAL1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Adhesion molecule-like X-linked, ADMLXHHA, anosmin-1, KAL, KALIG1, KALIG-1, Kallmann syndrome 1 sequence, Kallmann syndrome interval gene 1, Kallmann syndrome protein, Kallmann syndrome-1 sequence (anosmin-1), KMS
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human Anosmin. The immunogen is located within amino acids 430 - 480 of Anosmin. Amino Acid Squence: KVYWKKTEDPTVNRYHVR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Endocrinology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3730
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.