missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76580
This item is not returnable.
View return policy
Description
KAL1 Polyclonal specifically detects KAL1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| KAL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Adhesion molecule-like X-linked, ADMLXHHA, anosmin-1, KAL, KALIG1, KALIG-1, Kallmann syndrome 1 sequence, Kallmann syndrome interval gene 1, Kallmann syndrome protein, Kallmann syndrome-1 sequence (anosmin-1), KMS | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3730 | |
| Store at 4°C. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 ug/ml | |
| NP_000207 | |
| ANOS1 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human Anosmin. The immunogen is located within amino acids 430 - 480 of Anosmin. Amino Acid Squence: KVYWKKTEDPTVNRYHVR | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction