missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kaiso Rabbit anti-Human, Mouse, Rat, Clone: 10O4V2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16272-100UL
This item is not returnable.
View return policy
Description
Kaiso Monoclonal antibody specifically detects Kaiso in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
| Kaiso | |
| Monoclonal | |
| Unconjugated | |
| KAISOkaiso transcription factor, transcriptional regulator Kaiso, WUGSC:H_DJ525N14.1, zinc finger and BTB domain containing 33, Zinc finger and BTB domain-containing protein 33, ZNF348kaiso, ZNF-kaiso | |
| A synthetic peptide corresponding to a sequence within amino acids 573-672 of human Kaiso (Q86T24). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| 10O4V2 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10009 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction