Learn More
Tocris Bioscience™ Kainic acid
Kainate agonist; excitant and neurotoxin
Specifications
| Assay Percent Range | >98% |
|---|---|
| Chemical Name or Material | (2 S,3 S,4 S)-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic acid |
| CAS | 487-79-6 |
Description
Kainic acid, also known as kainate, is a selective agonist of kainate ionotropic glutamate receptors (EC50 = 0.6 - 7.4 μM) and a partial agonist at AMPA receptors (EC50 = 31 μM - 170 μM). Kainic acid is used to model epilepsy in vivo and to study the mechanisms of neurodegeneration and neurocytosis induced by excess stimulation by kainic acid. Kainate is shown to be involved in amyloidogenic processing of amyloid precursor protein and Aβ peptides in Alzheimer's disease.Related compounds include synthetic Kainic acid (Cat. No. 7065), the kainate receptor agonist Domoic acid (Cat. No. 0269) and the antagonist NBQX (Cat. No. 0373).
Specifications
| >98% | |
| (2 S,3 S,4 S)-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic acid | |
| 487-79-6 |
Safety and Handling
Recommended Storage : Store at Room Temperature
For Research Use Only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.