missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Kainic acid (synthetic)

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
Kainic acid, also known as kainate, is a selective agonist of kainate ionotropic glutamate receptors (EC50 = 0.6 μM - 7.4 μM) and a partial agonist at AMPA receptors (EC50 = 31 μM - 170 μM). Kainic acid is used to model epilepsy in vivo and to study the mechanisms of neurodegeneration and neurocytosis induced by excess stimulation by kainic acid. Kainate is shown to be involved in amyloidogenic processing of amyloid precursor protein and Aβ peptides in Alzheimer's. Synthetic version of Kainic acid (Cat. No. 0222)
Related compounds include the kainate receptor agonist Domoic acid (Cat. No. 0269) and the antagonist NBQX (Cat. No. 0373).
Specifications
Specifications
| Chemical Name or Material | (2S,3S,4S)-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic acid |
| CAS | 487-79-6 |
| Quantity | 50 mg |
| Molecular Formula | C10H15NO4 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction