missing translation for 'onlineSavingsMsg'
Learn More

Kaempferide, MedChemExpress

Product Code. 16456528
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456528

Brand: Medchem Express HY15449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kaempferide is an O-methylated flavonol, a type of chemical compound.

Kaempferide is an O-methylated flavonol, a type of chemical compound.

Specifications

Chemical Name or Material Kaempferide
Physical Form Solid
CAS 491-54-3
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H12O6
For Use With (Application) Cancer-programmed cell death
Synonym Kaempferol 4'-O-methyl ether
Solubility Information DMSO : 20 mg/mL (66.61 mM; Need ultrasonic)
SMILES O=C1C(O)=C(C2=CC=C(OC)C=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 300.26
Formula Weight 300.26
Percent Purity 99.42%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.