missing translation for 'onlineSavingsMsg'
Learn More

KAD-1229 Calcium Hydrate, TRC

Product Code. 30395327
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395327 Supplier Toronto Research Chemicals Supplier No. TRCK09000050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material KAD-1229 Calcium Hydrate
CAS 207844-01-7
Molecular Formula 2 C19 H24 N O3 . Ca . 2 H2 O
Quantity 50 mg
Synonym 2H-Isoindole-2-butanoic acid, octahydro-γ-oxo-α-(phenylmethyl)-, calcium salt, hydrate (2:1:2), (αS,3aR,7aS)-, 2H-Isoindole-2-butanoic acid, octahydro-γ-oxo-α-(phenylmethyl)-, calcium salt, dihydrate, (αS,3aR,7aS)- (9CI), (S)-2-Benzyl-4-oxo-4-(cis-perhydroisoindol-2-yl)butyric acid calcium salt dihydrate, Mitiglinide calcium dihydrate
InChI Formula InChI=1S/2C19H25NO3.Ca.2H2O/c2*21-18(20-12-15-8-4-5-9-16(15)13-20)11-17(19(22)23)10-14-6-2-1-3-7-14;;;/h2*1-3,6-7,15-17H,4-5,8-13H2,(H,22,23);;2*1H2/q;;+2;;/p-2/t2*15-,16+,17-;;;/m00.../s1
SMILES O.O.[Ca+2].[O-]C(=O)[C@H](CC(=O)N1C[C@H]2CCCC[C@H]2C1)Cc3ccccc3.[O-]C(=O)[C@H](CC(=O)N4C[C@H]5CCCC[C@H]5C4)Cc6ccccc6
IUPAC Name calcium;(2S)-4-[(3aS,7aR)-1,3,3a,4,5,6,7,7a-octahydroisoindol-2-yl]-2-benzyl-4-oxobutanoate;dihydrate
Molecular Weight (g/mol) 704.91
Formula Weight 704.335
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.