missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KAAG1 Recombinant Human Monoclonal Antibody (2H3)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560722
This item is not returnable.
View return policy
Description
KAAG1 Recombinant Monoclonal Antibody for ELISA
Specifications
| KAAG1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UBP8 | |
| KAAG1 | |
| Recombinant Human KAAG1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 2H3 | |
| Unconjugated | |
| KAAG1 | |
| KAAG1; kidney associated antigen 1; kidney-associated antigen 1; RU2 antisense gene protein; RU2AS | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 353219 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction