Learn More
Invitrogen™ KAAG1 Biosimilar Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558965
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term ADCT-901.
KAAG1 (Kidney-associated antigen 1) is a gene implicated in various kidney functions and has been particularly associated with renal cell carcinoma. It is expressed predominantly in the kidney, and its role in cancer biology includes its association with cellular proliferation and survival. The structural composition of KAAG1 includes characteristic motifs that enable its interaction with other cellular proteins, influencing pathways related to cell growth and death. Research has demonstrated that KAAG1 can be a valuable marker and potential therapeutic target for kidney cancer due to its significant involvement in the progression of the disease. Further studies are warranted to understand its complete biological role and therapeutic potential.
Specifications
| KAAG1 Biosimilar | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.