missing translation for 'onlineSavingsMsg'
Learn More

KA2/GRIK5/Glutamate Receptor KA2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18427052 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427052 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427052 Supplier Novus Biologicals Supplier No. NBP176853MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KA2/GRIK5/Glutamate Receptor KA2 Polyclonal antibody specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen KA2/GRIK5/Glutamate Receptor KA2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EAA2, Excitatory amino acid receptor 2, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, KA2GRIK2
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human Grik5. The immunogen is located within the last 50 amino acids of Grik5. Amino Acid Squence: EATSPPRPRPGPAGPRE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2901
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.