missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KA2/GRIK5/Glutamate Receptor KA2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£141.00
Specifications
| Antigen | KA2/GRIK5/Glutamate Receptor KA2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 - 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
KA2/GRIK5/Glutamate Receptor KA2 Polyclonal specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| KA2/GRIK5/Glutamate Receptor KA2 | |
| Western Blot 0.5 - 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2-5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_002079 | |
| 2901 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human Grik5. The immunogen is located within the last 50 amino acids of Grik5. Amino Acid Squence: EATSPPRPRPGPAGPRE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| EAA2, Excitatory amino acid receptor 2, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, KA2GRIK2 | |
| GRIK5 | |
| IgG | |
| Affinity Purified | |
| This Grik5 antibody does not cross-react with Grik4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title