missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KA1/GRIK4/Glutamate Receptor KA1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76852F
This item is not returnable.
View return policy
Description
KA1/GRIK4/Glutamate Receptor KA1 Polyclonal antibody specifically detects KA1/GRIK4/Glutamate Receptor KA1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| KA1/GRIK4/Glutamate Receptor KA1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| EAA1, Excitatory amino acid receptor 1, glutamate receptor KA1, Glutamate receptor KA-1, glutamate receptor, ionotropic kainate 4, glutamate receptor, ionotropic, kainate 4, KA1GRIK | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of the human Grik4. The immunogen is located within amino acids 30 - 80 of Grik4. Amino Acid Squence: RINRAPERLGKAK | |
| 0.1 mL | |
| Neuronal Cell Markers, Neurotransmission | |
| 2900 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction