missing translation for 'onlineSavingsMsg'
Learn More
Learn More
K15 (a putative transmembrane protein similar to LMP2A) Antibody (IF3/A10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50341CL1
This item is not returnable.
View return policy
Description
K15 (a putative transmembrane protein similar to LMP2A) Monoclonal antibody specifically detects K15 (a putative transmembrane protein similar to LMP2A) in Human,Virus samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| K15 (a putative transmembrane protein similar to LMP2A) | |
| Monoclonal | |
| CoraFluor 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human, Virus | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| IF3/A10 | |
| PBS | |
| Recombinant protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction