missing translation for 'onlineSavingsMsg'
Learn More

K15 (a putative transmembrane protein similar to LMP2A) Antibody (IF3/A10), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-50341AF488

Product Code. 18111378

  • £360.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

K15 (a putative transmembrane protein similar to LMP2A) Monoclonal specifically detects K15 (a putative transmembrane protein similar to LMP2A) in Human, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

K15 (a putative transmembrane protein similar to LMP2A)
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Recombinant protein
0.1 mL
Apoptosis, Virology Bacteria and Parasites
Human, Virus
Purified
Western Blot, ELISA, Immunohistochemistry
IF3/A10
Western Blot, ELISA, Immunohistochemistry
Mouse
Protein A purified
RUO
Primary
Store at 4C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.