missing translation for 'onlineSavingsMsg'
Learn More
Learn More
K15 (a putative transmembrane protein similar to LMP2A) Antibody (3B5/D7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50340UV
This item is not returnable.
View return policy
Description
K15 (a putative transmembrane protein similar to LMP2A) Monoclonal specifically detects K15 (a putative transmembrane protein similar to LMP2A) in Human, Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| K15 (a putative transmembrane protein similar to LMP2A) | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Recombinant protein | |
| 0.1 mL | |
| Apoptosis, Virology Bacteria and Parasites | |
| Human, Virus | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3B5/D7 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction