missing translation for 'onlineSavingsMsg'
Learn More
Learn More
K02288, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
K02288 is a potent bone morphogenetic protein (BMP) type I receptor inhibitor with IC50s of 1.8, 1.1, 6.4 nM for ALK1, ALK2 and ALK6, respectively. K02288 shows slightly weaker inhibition against ALK3 and ALK6 with IC50s of of 5-34 nM.
Specifications
Specifications
| Chemical Name or Material | K02288 |
| CAS | 1431985-92-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H20N2O4 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 58.6 mg/mL (166.30 mM) |
| SMILES | OC1=CC=CC(C2=CC(C3=CC(OC)=C(OC)C(OC)=C3)=C(N)N=C2)=C1 |
| Molecular Weight (g/mol) | 352.38 |
| Formula Weight | 352.38 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction