missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ K-UB-90 Combination Underbench Safety Storage Cabinets

Product Code. 15536865 Shop All asecos Products
Change view
Click to view available options
Configuration:
1 75 kg Insert, 1 10 L Sump, 1 x 25 kg Pull-out Shelf with PE-sump
1 75 kg Insert, 117 L Sump, 1 25 kg Pull-out Shelf with PE-sump
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Unit Size
15536865 1 75 kg Insert, 1 10 L Sump, 1 x 25 kg Pull-out Shelf with PE-sump Each
15654984 1 75 kg Insert, 117 L Sump, 1 25 kg Pull-out Shelf with PE-sump Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15536865 Supplier Asecos™ Supplier No. 3036500133844
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the safe, separated and ventilated storage of slightly aggressive and flammable hazardous substances in working areas.

  • Separated storage compartments with vertical dividing wall
  • Even and separate ventilation of each storage compartment via one central air duct (DN 75) ready for connection to a technical exhaust system
  • Fits perfectly under fume cupboards and work benches to save space
  • In accordance with EN 14470-1 and TRGS 510
  • Uniform appearance – no motley visual appearance due to storage in one cabinet
  • Can be provided with pull-out shelves, wing doors and drawers, height-adjustable shelves and bottom collecting sumps
  • Fire resistance of 90 minutes
  • Robust construction and longevity with body made of powder-coated sheet steel
  • User-friendly and safe – door and drawer remain open in any position, interior of the cabinet completely visible, access to all containers, 93° opening of wing doors
  • Safe – drawer fitted with earthing wire including clamp as standard, seal welded sump, drawers are self-closing in event of fire
  • No unauthorized use – door and drawer lockable with cylinder lock and locking state indicator (red/green)
  • Mobile – castors with plinth optionally available for easy moving of the cabinet

Specifications

Certifications/Compliance EN 14470-1, TRGS 510
Configuration 1 75 kg Insert, 1 10 L Sump, 1 x 25 kg Pull-out Shelf with PE-sump
Description Insert 75 kg, Pull-out shelf: 25 kg/11 L, Sump: 10 L, Max load: 300 kg
Dimensions (D x W x H) 502 x 1102 x 601 mm
Door Style Wing
For Use With (Application) Storage of slightly aggressive, flammable hazardous substances
Load Bearing Capacity 300 kg
No. of Doors 2
Surface Coating Powder-coated
Weight (Metric) 152.5 kg
Color Light Gray
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.