missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90

Product Code. 15411435 Shop All asecos Products
Change view
Click to view available options
Model:
K90.060.110.050.UB.2T
K90.060.110.050.UB.ST
K90.060.140.050.UB.3T
K90.060.140.050.UB.ST
Weight (Metric):
152.50 kg
186.50 kg
190.50 kg
196.50 kg
No. of Doors:
1
2
3
No. of Shelves:
1
2
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Weight (Metric) No. of Doors No. of Shelves unitSize
15411435 K90.060.110.050.UB.2T 152.50 kg 2 1 Each
15914366 K90.060.110.050.UB.ST 152.50 kg 1 1 Each
15984366 K90.060.140.050.UB.ST 186.50 kg 1 1 Each
15914376 K90.060.140.050.UB.3T 196.50 kg 3 2 Each
15184599 K90.060.140.050.UB.3T 190.50 kg 3 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy

For indoor use only

Product Code. 15411435 Supplier asecos™ Supplier No. 3036500130366
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of aggressive and flammable hazardous substances in working areas

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly and safe: door and drawer remain open in any position, interior of the cabinet completely visible — access to all containers, 93° opening of the wing door
  • Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: door and drawer lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system, extraction of both compartments through one ventilation connection
  • Certificates
    • Fire resistance 90 minutes
    • Type tested in accordance with EN 14470-1
    • EN 16121/16122 conformity
    • CE compliant

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Configuration 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), 1 x pull-out shelf (panel melamine resin-coated/PP)
Depth (Metric) Exterior 502 mm
Depth (Metric) Interior 380 mm
Dimensions (D x W x H) Interior Left: 380 x 470 x 502 mm, Right: 468 x 440 x 562 mm
Dimensions (D x W x H) 502 x 1102 x 601 mm
Door Style Wing, Right-hinged
For Use With (Application) Safe and approved storage of aggressive and flammable hazardous substances in working areas
Height (Metric) Exterior 601 mm
Height (Metric) Interior 502 mm
Model K90.060.110.050.UB.2T
No. of Doors 2
No. of Shelves 1
Shelf Type Pull-out
Surface Coating Powder-coated plain
Weight (Metric) 152.50 kg
Width (Metric) Exterior 1102 mm
Width (Metric) Interior 470 mm
Color Light Gray
Depth (English) Exterior 19.76 in.
Height (English) Exterior 23.66 in.
Height (Metric) Opening 502 mm
Material Steel
Product Type Combi Safety Storage Cabinet
Width (English) Exterior 43.39 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.