missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ K Type Extension Cables

Use these extension cables for a variety of applications. Cole-Parmer™ K Type Extension Cables feature 20-gauge (0.032 in. dia) Thermocouple Wire with PVC/PVC Insulation.

Brand:  Cole-Parmer™ 08516-35

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11760979

  • £51.20 / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket

Description

Description

  • Each probe comes with color coded ANSI mini-connectors: type J-black, type K-yellow and type T-blue
Specifications

Specifications

ANSI
Extension Cable
7.5 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers