missing translation for 'onlineSavingsMsg'
Learn More

K-Ras-IN-1, MedChemExpress

Product Code. 16480048
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480048 5 mg 5mg
16423597 50 mg 50mg
16417578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16480048 Supplier Medchem Express Supplier No. HY18674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress K-Ras-IN-1 is a K-Ras inhibitor. K-Ras-IN-1 binds to K-Ras (WT), K-Ras (G12D), K-Ras (G12V), and H-Ras. K-Ras-IN-1 has potential for the research of pancreatic, colon and lung carcinomas.

K-Ras-IN-1 is a K-Ras inhibitor. K-Ras-IN-1 binds to K-Ras (WT), K-Ras (G12D), K-Ras (G12V), and H-Ras. K-Ras-IN-1 has potential for the research of pancreatic, colon and lung carcinomas.

Specifications

Chemical Name or Material K-Ras-IN-1
CAS 84783-01-7
Color White
Purity Grade Notes Research
Molecular Formula C11H13NOS
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (482.42 mM; Need ultrasonic)
SMILES S=C(C1=CC=CC=C1O)N2CCCC2
Molecular Weight (g/mol) 207.29
Formula Weight 207.29
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.