missing translation for 'onlineSavingsMsg'
Learn More

K-Ras(G12C) inhibitor 12, MedChemExpress

Product Code. 16480068
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480068 5 mg 5mg
16447598 10 mg 10mg
16433617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16480068 Supplier Medchem Express Supplier No. HY18707
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress K-Ras(G12C) inhibitor 12 is a K-Ras(G12C) inhibitor, the half-maximum effective concentration (EC50) for K-Ras(G12C) inhibitor 12 in H1792 cells is 0.32 μM.

K-Ras(G12C) inhibitor 12 is a K-Ras(G12C) inhibitor, the half-maximum effective concentration (EC50) for K-Ras(G12C) inhibitor 12 in H1792 cells is 0.32 μM.

Specifications

Chemical Name or Material K-Ras(G12C) inhibitor 12
CAS 1469337-95-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H17ClIN3O3
Quantity 5 mg
Solubility Information DMSO : 16.67 mg/mL (37.07 mM; Need ultrasonic)
SMILES C=CC(N1CCN(C(CNC2=CC(I)=C(Cl)C=C2O)=O)CC1)=O
Molecular Weight (g/mol) 449.67
Formula Weight 449.67
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.