missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ K-cadherin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576343
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Cadherins constitute a family of transmembrane glycoproteins involved in Ca2+-dependent cell-cell interactions. The members of this family are differentially expressed in various tissues. They function in the maintenance of tissue integrity and morphogenesis. Cadherins are divided into type I and type II subgroups. Type I cadherins include epithelial cadherin (E-cadherin, cadherin-1 or uvomorulin), neural cadherin (N-cadherin or cadherin-2), placental cadherin (P-cadherin or cadherin-3) and retinal cadherin (R-cadherin or cadherin-4), whereas kidney cadherin (K-cadherin or cadherin-6) and osteoblast cadherin (OB-cadherin or cadherin-11) are type II cadherins. Cadherin-6 is expressed in the proximal tubule epithelium of the kidney from which most renal cell carcinomas are derived.
Specifications
| K-cadherin | |
| Polyclonal | |
| Unconjugated | |
| CDH6 | |
| CAD6; CADH6; cadherin 6; cadherin 6, type 2, K-cadherin; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin6; cadherin-6; Cdh6; k cadherin; KCAD; K-cadherin; Kidney cadherin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1004, 12563, 25409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55280, P55285, P97326 | |
| CDH6 | |
| Recombinant protein of human K-cadherin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction