missing translation for 'onlineSavingsMsg'
Learn More

K-252a, 98%, , Thermo Scientific Chemicals

Product Code. 10018751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10018751 1 mg 1mg
10246021 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10018751 Supplier Thermo Scientific Acros Supplier No. 328310010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97161-97-2
Molecular Formula C27H21N3O5
Molecular Weight (g/mol) 467.5
MDL Number MFCD00161522
InChI Key KOZFSFOOLUUIGY-UHFFFAOYSA-N
Synonym methyl 6-hydroxy-5-methyl-13-oxo-5,6,7,8,14,15-hexahydro-13h-5,8-epoxy-4b,8a,14-triazadibenzo b,h cycloocta 1,2,3,4-jkl cyclopenta e-as-indacene-6-carboxylate
PubChem CID 3813
SMILES CC12C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)(C(=O)OC)O

Specifications

Assay Percent Range 97.5% min. (HPLC)
Quantity 1 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in dichloromethane,dmso
Formula Weight 467.5
Percent Purity 98%
Chemical Name or Material K-252a

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.