missing translation for 'onlineSavingsMsg'
Learn More

K 01-162, MedChemExpress

Product Code. 16458687
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458687 5 mg 5mg
16426088 10 mg 10mg
16412227 50 mg 50mg
16411419 1 mg 1mg
16406486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16458687 Supplier Medchem Express Supplier No. HY14533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress K 01-162 (K162) binds and destabilizes AβO (β-amyloid), with an EC50 of 80 nM.

K 01-162 (K162) binds and destabilizes AβO (β-amyloid), with an EC50 of 80 nM.

Specifications

Chemical Name or Material K 01-162
CAS 677746-25-7
Color White
Purity Grade Notes Research
Molecular Formula C15H14BrN
Quantity 5 mg
Synonym K162
Solubility Information DMSO : 14.29 mg/mL (49.59 mM; Need ultrasonic)
SMILES BrC1=CC(CC2=C3C=CC(N(C)C)=C2)=C3C=C1
Molecular Weight (g/mol) 288.18
Formula Weight 288.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.