missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JZL 184
Description
Potent and selective MAGL inhibitor. Blocks hydrolysis of the endocannabinoid 2-arachidonyl glycerol (2-AG) in vivo in the mouse brain (IC50 = 8 nM). Potentiates depolarization-induced suppression of excitability in cerebellar Purkinje neurons. Exhibits >300-fold selectivity for MAGL over FAAH in vitro. Attenuates nociception in neuropathic and inflammatory pain models. Also reduces free fatty acid levels in primary tumors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 520.49 |
| Purity | >98% |
| Molecular Weight (g/mol) | 520.494 |
| Chemical Name or Material | JZL 184 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?