missing translation for 'onlineSavingsMsg'
Learn More

JZL 184, MedChemExpress

Product Code. 16486686
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486686 100 mg 100mg
16446398 10 mg 10mg
16432507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486686 Supplier Medchem Express Supplier No. HY15249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JZL 184 is a potent, selective and irreversible MAGL inhibitor that blocks 2-Arachidonoylglycerol (2-AG) hydrolysis in brain membranes (IC50 of 8 nM). JZL 184 displays >300-fold selectivity for MAGL over FAAH.

JZL 184 is a potent, selective and irreversible MAGL inhibitor that blocks 2-Arachidonoylglycerol (2-AG) hydrolysis in brain membranes (IC50 of 8 nM). JZL 184 displays >300-fold selectivity for MAGL over FAAH.

Specifications

Chemical Name or Material JZL 184
CAS 1101854-58-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H24N2O9
Quantity 100 mg
Solubility Information DMSO : ≥ 35 mg/mL (67.24 mM)
SMILES OC(C1=CC=C(OCO2)C2=C1)(C3CCN(C(OC4=CC=C([N+]([O-])=O)C=C4)=O)CC3)C5=CC(OCO6)=C6C=C5
Molecular Weight (g/mol) 520.49
Formula Weight 520.49
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.