missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JX 06

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
JX 06 is a potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor (IC50 values are 49, 101 and 313 nM for PDK1, PDK2 and PDK3, respectively). Exhibits selectivity for PDK1/2/3 over PDK4 and a panel of 323 other kinases. Suppresses A549 lung cancer cell proliferation in vitro. Reduces tumor volume of A549 xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | Bis(morpholinothiocarbonyl) disulfide |
| CAS | 729-46-4 |
| Quantity | 10 mg |
| Target | PDK-1 Inhibitors |
| Molecular Formula | C10H16N2O2S4 |
| Formula Weight | 324.51 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction