missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JWA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571372
This item is not returnable.
View return policy
Description
JWA Polyclonal Antibody for Western Blot
JWA protein expression is affected by vitamin A and may be associated with the cytoskeleton. A protein similar to JWA in rats may play a role in the regulation of cell differentiation by inhibiting the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport.
Specifications
| JWA | |
| Polyclonal | |
| Unconjugated | |
| Arl6ip5 | |
| 5930404D22Rik; addicsin; addiscin; ADP ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor GTPase 6 interacting protein 5; ADP-ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor-like 6 interacting protein 5; ADP-ribosylation factor-like protein 6-interacting protein 5; ADP-ribosylation-like factor 6 interacting protein 5; Aip5; Aip-5; ARL-6-interacting protein 5; Arl6ip5; AV001879; cytoskeleton related vitamin A responsive protein; cytoskeleton-related vitamin A-responsive protein; dermal papilla derived protein 11; dermal papilla-derived protein 11; DERP11; glutamate transporter EAAC1 interacting protein; glutamate transporter EAAC1-interacting protein; glutamate transporter EEAC1-associated protein; GTRAP3-18; hp22; HSPC127; JM5; jmx; Jwa; PRA1 domain family 3; PRA1 family protein 3; Pra2; Praf3; Prenylated Rab acceptor protein 2; protein JWa; putative MAPK activating protein PM27; putative MAPK-activating protein PM27; Yip6b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 65106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8R5J9 | |
| Arl6ip5 | |
| synthetic peptide directed towards the N terminal region of human JWA. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction