missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT3 Antibody (S180-41), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59319UV
This item is not returnable.
View return policy
Description
EAAT3 Monoclonal antibody specifically detects EAAT3 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
| EAAT3 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SLC1A1 | |
| Fusion protein amino acids 1-524 (full length) of human EAAT3 (Uniprot: P43005) | |
| 100 μL | |
| 6505 | |
| Human, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S180-41 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects 60kDa. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction