missing translation for 'onlineSavingsMsg'
Learn More

JW 55, MedChemExpress

Product Code. 16495938
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16495938

Brand: Medchem Express HY13968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JW 55 is a potent and selective β-catenin signaling pathway inhibitor, which functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). JW 55 decreases auto-PARsylation of TNKS1/2 in vitro with IC50s of 1.9 μM and 830 nM respectively.

JW 55 is a potent and selective β-catenin signaling pathway inhibitor, which functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2). JW 55 decreases auto-PARsylation of TNKS1/2 in vitro with IC50s of 1.9 μM and 830 nM respectively.

Specifications

Chemical Name or Material JW 55
CAS 664993-53-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H26N2O5
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (115.08 mM)
SMILES O=C(C1=CC=CO1)NC2=CC=C(C(NCC3(C4=CC=C(OC)C=C4)CCOCC3)=O)C=C2
Molecular Weight (g/mol) 434.48
Formula Weight 434.48
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.