missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Juvenile Hormone I, TRC
Specifications
Specifications
| Chemical Name or Material | Juvenile Hormone I |
| CAS | 13804-51-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H30O3 |
| Quantity | 2.5 mg |
| Synonym | [2R-[2α(2E,6E),3α]]-7-Ethyl-9-(3-ethyl-3-methyloxiranyl)-3-methyl-2,6-nonadienoic acid methyl ester, (2E,6E)-7-Ethyl-9-[(2R,3S)-3-ethyl-3-methyloxiranyl]-3-methyl-2,6-nonadienoic acid methyl ester, (E,E)-cis-10,11-Epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoic acid methyl ester, (10R,11S)-(+)-Juvenile hormone I, (10R,11S)-Juvenile hormone I, (E,E)-cis-Juvenile hormone I, 10,11-Epoxy-7-ethyl-3,11-dimethyl-trans,trans-2,6-tridecadienoic acid methyl ester, 10,11-Epoxy-7-ethyl-3,11-dimethyltrideca-2-cis-6-dienoic acid methyl ester, AI 3-33972, AY 22243, C18-Juvenile hormone, Cecropia juvenile hormone, JH I, Juvenile hormone 1, Methyl (2E,6E,10Z)-10,11-epoxy-3,11-dimethyl-7-ethyl-2,6-tridecadienoate, Methyl (E,E cis)-10,11-epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoate, Methyl (E,E)-3,11-dimethyl-7-ethyl-cis-10,11-epoxytrideca-2,6-dienoate, Methyl 10,11-epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoate, Methyl 10,11-epoxy-7-ethyl-3,11-dimethyl-trans,trans,cis-2,6-tridecadienoate, Methyl 12,14-dihomojuvenate, Methyl 3,11-dimethyl-7-ethyl-cis-10,11-oxido-trans,trans-2,6-tridecadienoate, Methyl 7-ethyl-3,11-dimethyl-10,11-epoxy-2,6-tridecadienoate, Methyl cis-10,11-epoxy-3,11-dimethyl-7-ethyl-trans,trans-2,6-tridecadienoate, Methyl cis-10,11-epoxy-7-ethyl-3,11-dimethyl-trans,trans-2,6-tridecadienoate, Methyl trans,trans,cis-10,11-epoxy-7-ethyl-3,11-dimethyltrideca-2,6-dienoate, Methyl-12-homojuvenate, cis/trans-Methyl 10,11-epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoate |
| InChI Formula | InChI=1S/C18H30O3/c1-6-15(11-12-16-18(4,7-2)21-16)10-8-9-14(3)13-17(19)20-5/h10,13,16H,6-9,11-12H2,1-5H3/b14-13+,15-10+/t16-,18+/m1/s1 |
| SMILES | Cl.Clc1n(C)c2c(n1)c3c(nccc3)cc2N |
| Molecular Weight (g/mol) | 294.43 |
| Formula Weight | 294.2195 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?