Learn More
Justrite™ I'm Easy Funnel
Product Code. 10061112
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Product Code. Material unitSize
10061112 Polypropylene Each
1 options
Product Code. 10061112 Supplier Justrite™ Supplier No. 11202Y
Only null left
Add to basket
Add to basket
Aids pouring into smaller openings
  • Easily attaches to 1 gal. or larger Type I safety cans
  • Large rounded bowl provides protection from splashing and spills
  • Innovative hinged design allow pouring or filling from can without removing funnel
  • Durable polypropylene holds up to heavy use
Specifications
Material Polypropylene
Height (Metric) 28.6 cm