missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Horizontal and Vertical Drum Vents
Description
- Choose brass, stainless steel or nonmetallic vent
- For petroleum-based liquids, use non-metallic or brass vents
- For chlorinated solvents, use nonmetallic vent with stainless-steel flame arrester
- For corrosives and general solvents, use stainless steel vents
Vertical Drum Vent:
- Has operating valve that opens automatically to prevent pressure buildup inside drum
Horizontal Drum Vent
- Has operating valve that can be opened manually to relieve internal pressure
Order Info
All vents fit 2 in. (5.1cm) bung. Cast Iron EL Fitting for use with 17-186A and 17-186B also available; see cat. no. 19018555.
Certifications
FM approved.
Specifications
Specifications
| Certifications/Compliance | FM |
| Product Type | Vertical Drum Vent |
| Description | Opens at 5 psig Internal Pressure |
| For Use With (Application) | Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup |
| For Use With (Equipment) | Vertical Drums |
| Material | Brass |
| Includes | Operating Valve |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?