missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Biohazard Waste Can

Product Code. 12903685
Change view
Click to view available options
Capacity (English):
6 gal.
10 gal.
Diameter (English) Outer:
12 in.
13.94 in.
14 in.
Height (English):
16 in.
18 in.
18.25 in.
Includes:
Foot-operated Cover
Foot-operated Cover, Soundguard
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Diameter (English) Outer Height (English) Includes unitSize
12903685 10 gal. 13.94 in. 18.25 in. Foot-operated Cover, Soundguard Each
12373823 10 gal. 14 in. 18 in. Foot-operated Cover Each
11934977 6 gal. 12 in. 16 in. Foot-operated Cover Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12903685 Supplier Justrite™ Supplier No. 05935

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Justrite Biohazard Waste Can is a sturdy, steel can used to store contaminated laundry as well as other regulated waste. Can features a leakproof construction with foot-operated, self-closing lid.

Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.

  • Features leakproof, sturdy steel construction with foot-operated, self-closing lid
  • Eliminates any reason to reach into the waste area or touch the can
  • Holds biohazard bag (sold separately) securely around circumference of can top
  • Large biohazard label clearly identifies the contents and alerts workers

Not to be used for disposal of sharp items.

Certifications

FM approved

Compliance

Complies with OSHA 29CFR1910.1030.

Specifications

Certifications/Compliance FM, TUV, OSHA
Diameter (Metric) Outer 35.4 cm
Diameter (English) Outer 13.94 in.
Capacity (English) 10 gal.
Color White
Height (Metric) 46.4 cm
Includes Foot-operated Cover, Soundguard
For Use With (Application) Safe, temporary collection of biological waste
Height (English) 18.25 in.
Material Metal
Product Type Biohazard Waste Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.