missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Flame Arresters for Nonmetallic Liquid Disposal Safety Cans
Description
- Precision mesh construction
- Available in stainless steel
Specifications
Specifications
| Product Type | Replacement Flame Arrester |
| Color | Silver |
| For Use With (Equipment) | For Nonmetallic Liquid Disposal Safety Cans |
| Material | Stainless Steel |
| Width (English) | 3.5 in. |
| Height (English) | .625 in. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?