missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ EcoPolyBlend™ Spill Control Pallets
Made of up to 100% recycled polyethylene
£324.00 - £467.00
Specifications
Certifications/Compliance | SPCC, EPA, NFPA, U (NFPA), meets fire codes |
---|---|
Material | Polyethylene |
Type | Ecopolyblend Spill Control Pallet |
For Use With (Application) | Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs, protect against groundwater contamination |
For Use With (Equipment) | Drums |
Includes: Two and four drum square pallets include wide forklift pockets.
Product Code | Brand | Capacity (Metric) | Color | Height (Metric) | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Height (Metric) | Includes | Price | Quantity & Availability | ||
11919385
|
Justrite™
28623 |
250 L | Black | 45.7 cm | Two drums |
£324.00
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||
11939385
|
Justrite™
28635 |
276 L | Black | 26 cm | 4-Drum Square |
£467.00
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
||
11949385
|
Justrite™
28631 |
28.4 L | Black | 22.9 cm | 4 Drum In-Line |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11929385
|
Justrite™
28627 |
28.4 L | Black | 29.5 cm | 3 drums |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Beschreibung
- Protect against groundwater contamination, using post-industrial material
- Robust pallets withstand the toughest environments for superior spill control
- Durable, UV-protected polyethylene is nearly impervious to chemical attack from hazardous acids, caustics, and solvents
- Seamless construction eliminates leaks—even if a 55-gallon (208L) drum fails and completely empties
- Oversized leakproof sumps meet or exceed regulations
- Sturdy, removable 2.5 in. (64mm) thick grates offer superior crack resistance; self-locating
- Black pallets constructed of 100% recycled content; dark color camouflages dirt and keeps work places looking clean
- Yellow pallets ideal for high visibility or liquid segregation; virgin poly sumps with 100% recycled grates (a total of 35% recycled content)
- Square pallet may be equipped with an optional ramp
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
Compliance
Meet or exceed EPA 40 CFR 264.175; NFPA 1 the Uniform Fire Code 60.3.2.10; International Fire Code 2704.2.3; Spill Prevention, Control and Countermeasures (SPCC) Compliant; NFPA 30 9.13.1 and OSHA 29 CFR 1910.106(e)(2)(iii).
Spezifikation
SPCC, EPA, NFPA, U (NFPA), meets fire codes | |
Ecopolyblend Spill Control Pallet | |
Drums |
Polyethylene | |
Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs, protect against groundwater contamination | |
Chemical |