Learn More
3M™ Jupiter™ Calibration and Air Flow Indicator, 021-14-12P
Product Code. 15959276
Click to view available options
:
CE Approved
Product Code. 15959276
Brand: 3M™ 7000104088
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Jupiter™ Calibration and Air Flow Indicator enables the checking and calibration of 3M™ Jupiter™ Powered Air Turbo.
  • Ensure 3M™ Jupiter™ Powered Air Turbo is exceeding its minimum air flow rate
  • Ensure powered air turbo alarm point is correctly set
  • Air flow must be checked before using powered air turbo
  • Powered Air Turbo must be calibrated once a month
  • Accessory