Learn More
3M™ Jupiter™ Air-Fed Powered Turbo Respirators
Provides respiratory protection against particulate and gaseous contaminants.
Brand: 3M™ 7100003547
Additional Details : Weight : 1.34100kg
Description
- Decontamination comfort belt for improved fit and easier cleaning
- Option of five filter types to select from, depending on work environment
- Modern, ergonomic design
- Can be worn during shower decontamination
- Fully compatible with all 3M™ Versaflo™ Headtops and Breathing Tubes
- Integrated acoustic and visual alarm indicates if airflow to the headtop is reduced or battery charge runs low
Specifications
4 to 8 hours | |
CE Approved | |
Blue, Black | |
Used to Provides Protection Against Particulates, Gas and Vapors | |
0850010P |
NiMH | |
EN 12941 TH2, EN 12941 TH3 | |
PAPR Assembly, No Headtop | |
Turbo unit only: motor, fan, comfort belt and instructions; helmet and hose not included | |
1 Ea. |