Learn More
Invitrogen™ JunD Recombinant Superclonal™ Antibody (2HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710701
Description
This antibody is predicted to react with monkey, sheep, mouse, rat, zebra fish and bovine based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
Specifications
| JunD | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P17535 | |
| JUND | |
| Peptide, multiple (2-16, 132-149, 299-313). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 2HCLC | |
| Unconjugated | |
| JUND | |
| activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3727 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.