missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunD Rabbit anti-Human, Mouse, Rat, Clone: 5L7Y3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | JunD |
|---|---|
| Clone | 5L7Y3 |
| Applications | ChIP Assay, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18383153
|
Bio-Techne
NBP3-16600-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18390503
|
Novus Biologicals
NBP3-16600-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
JunD Monoclonal antibody specifically detects JunD in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, ImmunoprecipitationSpecifikationer
| JunD | |
| ChIP Assay, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human JunD (P17535). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5L7Y3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 3727 | |
| IgG | |
| Affinity purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel