missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunD Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22439JF525
This item is not returnable.
View return policy
Description
JunD Polyclonal antibody specifically detects JunD in Mouse samples. It is validated for Flow Cytometry,Western Blot,ChIP assay (ChIP),Immunocytochemistry/ Immunofluorescence
Specifications
| JunD | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Western Blot, ChIP Assay, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 525 | |
| activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D | |
| Synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD. | |
| 0.2 mL | |
| Phospho Specific | |
| 3727 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction