missing translation for 'onlineSavingsMsg'
Learn More

JunD Antibody - BSA Free, Novus Biologicals™

Product Code. 18404502 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
50 ug
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404502 50 ug 50µg
18190472 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18404502 Supplier Novus Biologicals Supplier No. NBP22243950ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

JunD Polyclonal specifically detects JunD in Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JunD
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ChIP Assay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10 ug/ml, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunocytochemistry/Immunofluorescence 2 - 3 ug/ml, Chromatin Immunoprecipitation (ChIP) 1-3 ul
Formulation PBS with 0.05% Sodium Azide
Gene Alias activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D
Gene Symbols JUND
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD.
Purification Method Immunogen affinity purified
Quantity 50 ug
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3727
Reconstitution Reconstitute with 0.2 ml PBS to a final concentration of 1 mg/ml.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.