missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunB/AP-1 Antibody (5H4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26185-100ul
This item is not returnable.
View return policy
Description
JunB/AP-1 Monoclonal antibody specifically detects JunB/AP-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| JunB/AP-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry | |
| 5H4 | |
| ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B | |
| A synthesized peptide derived from Human JunB/AP-1 [UniProt P17275] | |
| 100 μL | |
| Phospho Specific | |
| 3726 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction